Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diphenyl-1,4-benzoquinone 99.0+%, TCI America™
SDP

Catalog No. D116625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D116625G 25 g
D11665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D116625G Supplier TCI America Supplier No. D116625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 844-51-9
Molecular Formula C18H12O2
Molecular Weight (g/mol) 260.29
MDL Number MFCD00001600
InChI Key QYXHDJJYVDLECA-UHFFFAOYSA-N
Synonym 2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
PubChem CID 70055
IUPAC Name 2,5-diphenylcyclohexa-2,5-diene-1,4-dione
SMILES O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 219°C
Quantity 25 g
Formula Weight 260.29
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diphenyl-1,4-benzoquinone
EINECSNumber (4)-0049
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.