Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diphenyl-p-benzoquinone, 96%

Catalog No. AAA1536914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1536914 25 g
AAA1536906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1536914 Supplier Thermo Scientific Chemicals Supplier No. A1536914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 844-51-9 | C18H12O2 | 260.29 g/mol

2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

Solubility
Soluble in hot Toluene (almost transparency).

Notes
Incompatible with Oxidizing agents, Reducing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 844-51-9
Molecular Formula C18H12O2
Molecular Weight (g/mol) 260.29
MDL Number MFCD00001600
InChI Key QYXHDJJYVDLECA-UHFFFAOYSA-N
Synonym 2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
PubChem CID 70055
IUPAC Name 2,5-diphenylcyclohexa-2,5-diene-1,4-dione
SMILES O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 215°C to 219°C
Quantity 25 g
Beilstein 2051812
Solubility Information Soluble in hot Toluene (almost transparency).
Formula Weight 260.3
Percent Purity 96%
Chemical Name or Material 2,5-Diphenyl-p-benzoquinone
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 212-680-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.