Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diphenylfuran 98.0+%, TCI America™
SDP

Catalog No. D31761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31761G Supplier TCI America Supplier No. D31761G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 955-83-9
Molecular Formula C16H12O
Molecular Weight (g/mol) 220.27
MDL Number MFCD00037353
InChI Key VUPDHIIPAKIKAB-UHFFFAOYSA-N
PubChem CID 70387
ChEBI CHEBI:50459
IUPAC Name 2,5-diphenylfuran
SMILES O1C(=CC=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 90°C
Color White-Yellow
Boiling Point 345°C
Quantity 1 g
Formula Weight 220.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diphenylfuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.