Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-diphenyloxazole | 92-71-7 | MFCD00005306 | 221.26 g·mol⁻¹ | C15H11NO | 5 G
SDP

Catalog No. 5000303814
Encompass_Preferred

2,5-Diphenyloxazole is an organic fluorescent dye used as a primary scintillator and wavelength shifter in liquid and solid scintillation applications and in fluorescence studies. It is supplied as a powder and is identified by CAS 92-71-7 for unambiguous reference.

  • Organic fluorescent dye suitable as a primary scintillator and wavelength shifter.
  • Useful for liquid and solid scintillation spectrometry and fluorescence applications.
  • Powder form for laboratory preparation and formulation.
  • Molecular formula C15H11NO; molecular weight 221.26 g·mol⁻¹.
  • CAS number 92-71-7 for clear chemical identification.

Catalog No. 50-003-03814 Supplier Medchemexpress LLC Supplier No. HYW0140185G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2,5-Diphenyloxazole is an organic fluorescent dye used as a primary scintillator and wavelength shifter in liquid and solid scintillation applications and in fluorescence studies. It is supplied as a powder and is identified by CAS 92-71-7 for unambiguous reference.

  • Organic fluorescent dye suitable as a primary scintillator and wavelength shifter.
  • Useful for liquid and solid scintillation spectrometry and fluorescence applications.
  • Powder form for laboratory preparation and formulation.
  • Molecular formula C15H11NO; molecular weight 221.26 g·mol⁻¹.
  • CAS number 92-71-7 for clear chemical identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.