Learn More
Medchemexpress LLC 2,5-diphenyloxazole | 92-71-7 | MFCD00005306 | 99.4% | 221.26 g·mol⁻¹ | C15H11NO | 1 G
SDP
Catalog No. 5000302089
Encompass_Preferred
2,5-Diphenyloxazole (PPO) is an organic fluorescent dye and primary scintillator used as a wavelength shifter; it absorbs high-energy photons and re-emits visible light, making it suitable for liquid scintillation, radiation detection, and fluorescence studies.
  • Acts as a primary scintillator and wavelength shifter for liquid scintillation and radiation detectors.
  • High purity (99.4%) suitable for analytical and scintillation-grade use.
  • Chemical formula C15H11NO, molecular weight 221.26 g·mol⁻¹.
  • Available in small pack sizes suitable for laboratory research and formulation into scintillation cocktails.
  • Stable under normal laboratory conditions when stored in a cool, dry place.
Catalog No. 50-003-02089 Supplier Medchemexpress LLC Supplier No. HYW0140181G
2,5-Diphenyloxazole (PPO) is an organic fluorescent dye and primary scintillator used as a wavelength shifter; it absorbs high-energy photons and re-emits visible light, making it suitable for liquid scintillation, radiation detection, and fluorescence studies.
  • Acts as a primary scintillator and wavelength shifter for liquid scintillation and radiation detectors.
  • High purity (99.4%) suitable for analytical and scintillation-grade use.
  • Chemical formula C15H11NO, molecular weight 221.26 g·mol⁻¹.
  • Available in small pack sizes suitable for laboratory research and formulation into scintillation cocktails.
  • Stable under normal laboratory conditions when stored in a cool, dry place.