Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diphenyloxazole, 99%, scintillation grade

Catalog No. AC117380025
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117380025 2.5 kg Plastic Drum
AC117380250 25 g Plastic Bottle
AC117381000 100 g Plastic Bottle
AC117385000 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC117380025 Supplier Thermo Scientific Chemicals Supplier No. 117380025
Only null left
Add to Cart

CAS: 92-71-7 | C15H11NO | 221.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for fluorescence/luminescence spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-71-7
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
MDL Number MFCD00005306
InChI Key CNRNYORZJGVOSY-UHFFFAOYSA-N
Synonym PPO
PubChem CID 7105
IUPAC Name 2,5-diphenyl-1,3-oxazole
SMILES O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C to 72°C
Physical Form Crystalline Powder
CAS Min % 98.5
CAS Max % 100.0
Background < 25 cpm
Density 1.06g/mL
Color White
Quantity 2.5 kg
Assay Percent Range 99%
Infrared Spectrum Authentic
Beilstein 27, 78
Packaging Plastic Drum
Formula Weight 221.26
Percent Purity ≥98.5%
Chemical Name or Material 2, 5-Diphenyloxazole, 99%
Show More Show Less
EINECSNumber 202-181-3
RTECSNumber RP6825000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.