Learn More
Cole-Parmer 2.5" FILLED SANITARY GAUGE 0-6
Supplier:  Cole-Parmer 6846056
Encompass_Preferred
Accuracy:±2.0% full-scale;Dial Size (in):2.5;Min Pressure (PSI):0; Max Pressure (PSI):60; Liquid Fill:Pharmaceutical/food quality USP-grade glycerin (99.5% pure). COLE PARMER mfg# EW-68460-56
Catalog No. NC1942444
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov