Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,5-Furandicarboxaldehyde

Catalog No. 7283731G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-837-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-837-31G Supplier Sigma Aldrich Supplier No. 7283731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,5-Furandicarboxaldehyde

Specifications

CAS 823-82-5
Molecular Formula C6H4O3
Linear Formula C6H4O3
MDL Number MFCD00671517
Quantity 1 g
Synonym 2,5-Diformylfuran; 2,5-Furandicarbaldehyde; 5-Formylfurfural
Molecular Weight (g/mol) 124.09
Percent Purity ≥96.5% (HPLC); 97%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.