Learn More
Medchemexpress LLC HY-W002105 1g Medchemexpress, 2,5-Furandicarboxylic acid CAS:3238-40-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0021051G
Encompass_Preferred
Medchemexpress, HY-W002105 1g 2,5-Furandicarboxylic acid CAS:3238-40-2 2,5-Furandicarboxylic acid is a biomass-derived diacid that can be used to make polymers including polyethylene furandicarboxylate (PEF). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4470