Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Furandicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. F071025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F071025G 25 g
F07105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F071025G Supplier TCI America Supplier No. F071025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3238-40-2
Molecular Formula C6H4O5
Molecular Weight (g/mol) 156.09
MDL Number MFCD00016582
InChI Key CHTHALBTIRVDBM-UHFFFAOYSA-N
Synonym 2,5-furandicarboxylic acid, dehydromucic acid, furan-2,5-dicarbonsaeure, 2,5-furandicarboxylicacid, 2,5-furandicarboxylate, furane-alpha,alpha'-dicarboxylic acid, furan 2,5-dicarboxylic acid, furan-2,5-dicarboxylate, furane-.alpha.,.alpha.'-dicarboxylic acid, 2,5 furan dicarboxylic acid
PubChem CID 76720
ChEBI CHEBI:84212
IUPAC Name furan-2,5-dicarboxylic acid
SMILES OC(=O)C1=CC=C(O1)C(O)=O

Specifications

Melting Point 320°C
Color White-Yellow
Quantity 25 g
Formula Weight 156.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Furandicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.