Learn More
US Plastic Corporation 2-1/2 Gallon E-Z Fill Container
SDP
Catalog No. NC1067345
Encompass
Filling this container with chemicals is easy with a 63mm angled front opening. The contents of this jug can then be dispensed through the molded drain and spigot configuration.
Catalog No. NC1067345 Supplier US Plastic Corporation Supplier No. 65368
Filling this container with chemicals is easy with a 63mm angled front opening. The contents of this jug can then be dispensed through the molded drain and spigot configuration.