Learn More
US Plastic Corporation 2.5 GAL HIGH DENS PP LOW PROFI
SDP
Catalog No. NC9216625
Encompass
Carboy; 2-1/2 gal.; Large mouth opening; HDPE; Calibrated in gallons and liters; Lightweight; Commercial grade; For use with chemicals, pharmaceuticals, cosmetics, liquid foods, and others; 5% off; Capacity: 10L; 12/CS
Catalog No. NC9216625 Supplier US Plastic Corporation Supplier No. 72028
Carboy; 2-1/2 gal.; Large mouth opening; HDPE; Calibrated in gallons and liters; Lightweight; Commercial grade; For use with chemicals, pharmaceuticals, cosmetics, liquid foods, and others; 5% off; Capacity: 10L; 12/CS