Learn More
US Plastic Corporation 2.5 GALLON CARBOY HANDLE SPOUT
SDP
Catalog No. NC1724828
Encompass
2-1/2 Gallon Tamco® Modified Nalgene™ Wide Mouth LDPE Carboy with a Handle & SpoutLDPE CarboyPolypropylene capGraduated in gallons & liter100mm cap sizeDimensions: 250mm Dia. x 389mm Hgt.Meets FDA standards
Catalog No. NC1724828 Supplier US Plastic Corporation Supplier No. 67002
2-1/2 Gallon Tamco® Modified Nalgene™ Wide Mouth LDPE Carboy with a Handle & SpoutLDPE CarboyPolypropylene capGraduated in gallons & liter100mm cap sizeDimensions: 250mm Dia. x 389mm Hgt.Meets FDA standards