Learn More
Electron Microscopy Sciences 2.5% Glutaraldehyde in Sodium Cacodylate Buffer, 7.4 pH 10 ML
SDP
Catalog No. 5025941
Encompass_Preferred
Premixed, prebuffered fixatives, are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 ml glass vials (10 ml/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives. In Sodium Cacodylate Buffer.
pH: 7.4
Catalog No. 50-259-41 Supplier Electron Microscopy Sciences Supplier No. 15960
Premixed, prebuffered fixatives, are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 ml glass vials (10 ml/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives. In Sodium Cacodylate Buffer.
pH: 7.4