Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-hexanedione-d10 | 97135-07-4 | MFCD01074031 | 124.20 g·mol⁻1 | C6D10O2 | 100mg
SDP

Catalog No. 5000268220
Encompass_Preferred

2 5-Hexanedione-d10 is the deuterium labeled 2 5-Hexanedione[1]

Catalog No. 50-002-68220 Supplier Medchemexpress LLC Supplier No. HYY0315S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2 5-Hexanedione-d10 is the deuterium labeled 2 5-Hexanedione[1]

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.