Learn More
Ancare Corporation 2.5 INCH STRAIGHT BALL POINT D
SDP
Catalog No. NC9222132
Encompass
Drinking Spout, Ball Point; 2.5 in. L; Straight; Have two balls and a 5/16 ft. opening; Provide cleaner water by significantly reducing the backflow of bacteria and debris into the water bottle
Catalog No. NC9222132 Supplier Ancare Corporation Supplier No. TD100
Drinking Spout, Ball Point; 2.5 in. L; Straight; Have two balls and a 5/16 ft. opening; Provide cleaner water by significantly reducing the backflow of bacteria and debris into the water bottle