Learn More
Apexbio Technology LLC 2.5 mM dNTP Mixture 10x1ml
SDP
Catalog No. 5000090220
Encompass_Preferred
2 5 mM dNTP Mixture is a balanced solution of deoxynucleotide triphosphates (dATP dCTP dGTP and dTTP) commonly used as substrates for DNA polymerases during in vitro DNA synthesis By providing equimolar amounts of each nucleotide this mixture ensures accurate and efficient DNA replication and amplification minimizing errors due to imbalanced nucleotide concentrations It is widely employed in polymerase chain reaction (PCR) quantitative PCR DNA sequencing and other molecular biology techniques that require reliable DNA synthesis supporting precise genetic analysis and experimental reproducibility
Catalog No. 50-000-90220 Supplier Apexbio Technology LLC Supplier No. K104010
2 5 mM dNTP Mixture is a balanced solution of deoxynucleotide triphosphates (dATP dCTP dGTP and dTTP) commonly used as substrates for DNA polymerases during in vitro DNA synthesis By providing equimolar amounts of each nucleotide this mixture ensures accurate and efficient DNA replication and amplification minimizing errors due to imbalanced nucleotide concentrations It is widely employed in polymerase chain reaction (PCR) quantitative PCR DNA sequencing and other molecular biology techniques that require reliable DNA synthesis supporting precise genetic analysis and experimental reproducibility