Learn More
Cayman Chemical 2 5-nhydro-D-mnItol 50mg
SDP
Catalog No. 502986323
Encompass_Preferred
An antimetabolic fructose analogue that increases food intake in rats
Catalog No. 50-298-6323 Supplier Cayman Chemical Supplier No. 2167350 MG
An antimetabolic fructose analogue that increases food intake in rats