Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(5-nitro-1H-imidazol-1-yl)ethanol | 5006-68-8 | MFCD00159670 | 98.0% | 157.13 g/mol | C5H7N3O3 | 250 MG
SDP

Catalog No. 5000303623
Encompass_Preferred

Metronidazole impurity 1 (2-(5-nitro-1H-imidazol-1-yl)ethanol; CAS 5006-68-8) is a characterized impurity of metronidazole used as a reference standard and drug-intermediate in pharmaceutical research. The material is supplied for research use only and is provided with analytical documentation and handling guidance.

  • Chemical formula: C5H7N3O3.
  • Molecular weight: 157.13 g/mol.
  • Intended use: reference standard and drug-intermediate for analytical and formulation research.
  • Typical pack sizes: 100 mg, 250 mg, 500 mg, 1 g.
  • Storage recommendations: store as a solid at room temperature; in solvent store at -80°C or -20°C per stability guidance.
  • Accompanying data: analytical reports such as LC/MS, NMR, and HPLC are available.

Catalog No. 50-003-03623 Supplier Medchemexpress LLC Supplier No. 250MGHYW015509
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metronidazole impurity 1 (2-(5-nitro-1H-imidazol-1-yl)ethanol; CAS 5006-68-8) is a characterized impurity of metronidazole used as a reference standard and drug-intermediate in pharmaceutical research. The material is supplied for research use only and is provided with analytical documentation and handling guidance.

  • Chemical formula: C5H7N3O3.
  • Molecular weight: 157.13 g/mol.
  • Intended use: reference standard and drug-intermediate for analytical and formulation research.
  • Typical pack sizes: 100 mg, 250 mg, 500 mg, 1 g.
  • Storage recommendations: store as a solid at room temperature; in solvent store at -80°C or -20°C per stability guidance.
  • Accompanying data: analytical reports such as LC/MS, NMR, and HPLC are available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.