Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Pyridinedicarboxylic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
100-26-5
C7H5NO4
167.12
MFCD00006297
LVPMIMZXDYBCDF-UHFFFAOYSA-N
2,5-pyridinedicarboxylic acid,isocinchomeronic acid,2,5-pyridinedicarboxylicacid,unii-4k1kgr926x,pyridinedicarboxylic acid-2,5,chembl88972,pyridine 2,5-dicarboxylate,pubchem12991,2,5-dicarboxy-pyridine,6-carboxy-nicotinic acid
7493
CHEBI:46865
pyridine-2,5-dicarboxylic acid
C1=CC(=NC=C1C(=O)O)C(=O)O
100-26-5
C7H5NO4
167.12
MFCD00006297
LVPMIMZXDYBCDF-UHFFFAOYSA-N
2,5-pyridinedicarboxylic acid,isocinchomeronic acid,2,5-pyridinedicarboxylicacid,unii-4k1kgr926x,pyridinedicarboxylic acid-2,5,chembl88972,pyridine 2,5-dicarboxylate,pubchem12991,2,5-dicarboxy-pyridine,6-carboxy-nicotinic acid
7493
CHEBI:46865
pyridine-2,5-dicarboxylic acid
C1=CC(=NC=C1C(=O)O)C(=O)O
Melting Point 256°C
Color Yellow to Green
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H3N)(COOH)2
Beilstein 22, 153
Merck Index 15, 5204
Solubility Information Solubility in water: insoluble in cold water. Other solubilities: soluble in hot water,insoluble in alcohol,ether and benzene,slightly soluble in boiling alcohol
Formula Weight 167.12
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Pyridinedicarboxylic acid, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-834-2

TSCA : TSCA