Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Pyridinedicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. P055225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P055225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P055225G Supplier TCI America Supplier No. P055225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-26-5
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006297
InChI Key LVPMIMZXDYBCDF-UHFFFAOYSA-N
Synonym 2,5-pyridinedicarboxylic acid, isocinchomeronic acid, 2,5-pyridinedicarboxylicacid, unii-4k1kgr926x, pyridinedicarboxylic acid-2,5, chembl88972, pyridine 2,5-dicarboxylate, pubchem12991, 2,5-dicarboxy-pyridine, 6-carboxy-nicotinic acid
PubChem CID 7493
ChEBI CHEBI:46865
IUPAC Name pyridine-2,5-dicarboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)C(=O)O

Specifications

Melting Point 254°C
Quantity 25 g
Formula Weight 167.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Pyridinedicarboxylic Acid
EINECSNumber (9)-1063
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.