Learn More
Medchemexpress LLC 2,5-pyrrolidinedione, 1,1'-[1,2-ethanediylbis[oxy(1-oxo-3,1-propanediyl)oxy]]bis(9CI) | 65869-63-8 | 99.2% | 400.34 | C16H20N2O10 | 5 G
SDP
Catalog No. 5000203833
Encompass_Preferred
Bis-PEG2-NHS ester is a non-cleavable two-unit polyethylene glycol (PEG) linker designed for antibody-agent conjugation (ADC). It contains an N-hydroxysuccinimide (NHS) ester reactive group for efficient amine coupling and is supplied as a white to off-white solid.
  • Non-cleavable two-unit PEG linker
  • NHS-ester functional group for amine coupling
  • High purity suitable for conjugation chemistry
  • Available in multiple pack sizes, including 5 G
  • Powder storage: -20°C for up to 3 years
Catalog No. 50-002-03833 Supplier Medchemexpress LLC Supplier No. HY1300885G
Bis-PEG2-NHS ester is a non-cleavable two-unit polyethylene glycol (PEG) linker designed for antibody-agent conjugation (ADC). It contains an N-hydroxysuccinimide (NHS) ester reactive group for efficient amine coupling and is supplied as a white to off-white solid.
  • Non-cleavable two-unit PEG linker
  • NHS-ester functional group for amine coupling
  • High purity suitable for conjugation chemistry
  • Available in multiple pack sizes, including 5 G
  • Powder storage: -20°C for up to 3 years