Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-pyrrolidinedione, 1,1'-[1,2-ethanediylbis[oxy(1-oxo-3,1-propanediyl)oxy]]bis- (9CI) | 65869-63-8 | 99.2% | 400.34 g/mol | C16H20N2O10 | 250 MG
SDP

Catalog No. 5000203007
Encompass_Preferred

Bis-PEG2-NHS ester is a noncleavable two-unit polyethylene glycol (PEG) linker bearing N-hydroxysuccinimide (NHS) ester reactive groups at both ends, designed for antibody-agent conjugation via amine-coupling chemistry for research use.

  • Noncleavable two-unit PEG linker for antibody-agent conjugation.
  • N-hydroxysuccinimide (NHS) ester reactive groups for amine coupling.
  • High purity, 99.2%.
  • Molecular weight 400.34 g/mol.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation may be required.
  • Storage: powder at -20°C; in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in multiple pack sizes, including 250 MG.

Catalog No. 50-002-03007 Supplier Medchemexpress LLC Supplier No. HY130088250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG2-NHS ester is a noncleavable two-unit polyethylene glycol (PEG) linker bearing N-hydroxysuccinimide (NHS) ester reactive groups at both ends, designed for antibody-agent conjugation via amine-coupling chemistry for research use.

  • Noncleavable two-unit PEG linker for antibody-agent conjugation.
  • N-hydroxysuccinimide (NHS) ester reactive groups for amine coupling.
  • High purity, 99.2%.
  • Molecular weight 400.34 g/mol.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation may be required.
  • Storage: powder at -20°C; in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in multiple pack sizes, including 250 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.