Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-pyrrolidinedione, 1,1'-[oxybis[(1-oxo-3,1-propanediyl)oxy]]bis- (9CI) | 65869-64-9 | MFCD20226397 | 97.0% | 356.28 | C14H16N2O9 | 500 MG
SDP

Catalog No. 5000203008
Encompass_Preferred

Bis-PEG1-NHS ester is a non-cleavable 1-unit polyethylene glycol (PEG) linker designed for antibody-agent conjugation (ADC) research applications. It contains two N-hydroxysuccinimide (NHS) ester reactive groups that enable conjugation to primary amines on proteins or payload molecules, and is supplied with high purity for analytical and synthetic workflows.

  • Non-cleavable 1-unit PEG linker for antibody-agent conjugation.
  • Contains two NHS ester reactive groups for conjugation to primary amines.
  • Intended for research use in ADC linker synthesis and bioconjugation workflows.
  • Purity 97.0% by HPLC.
  • Available in a 500 MG pack size suitable for preparative work.

Catalog No. 50-002-03008 Supplier Medchemexpress LLC Supplier No. HY130089500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG1-NHS ester is a non-cleavable 1-unit polyethylene glycol (PEG) linker designed for antibody-agent conjugation (ADC) research applications. It contains two N-hydroxysuccinimide (NHS) ester reactive groups that enable conjugation to primary amines on proteins or payload molecules, and is supplied with high purity for analytical and synthetic workflows.

  • Non-cleavable 1-unit PEG linker for antibody-agent conjugation.
  • Contains two NHS ester reactive groups for conjugation to primary amines.
  • Intended for research use in ADC linker synthesis and bioconjugation workflows.
  • Purity 97.0% by HPLC.
  • Available in a 500 MG pack size suitable for preparative work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.