Learn More
Medchemexpress LLC 2,5-pyrrolidinedione derivative (PEG1 NHS ester) | 173414-89-6 | 97.0% | 627.55 g/mol | C26H33N3O15 | 100 MG
SDP
Catalog No. 5000203062
Encompass_Preferred
m-C-tri(CH2-PEG1-NHS ester) is a non-cleavable, single-unit polyethylene glycol (PEG) antibody-drug conjugate (ADC) linker bearing an N-hydroxysuccinimide (NHS) ester for amine coupling. Supplied as a solid for research use, it provides a stable, non-cleavable spacer that improves solubility and facilitates conjugation.
  • Non-cleavable single-unit PEG linker for stable spacing
  • NHS ester functionality for efficient amine conjugation
  • Improves payload solubility
  • Supplied as a solid with ~97.0% purity (NMR)
  • Available in multiple sample sizes for research (50 mg-1 g)
Catalog No. 50-002-03062 Supplier Medchemexpress LLC Supplier No. HY44149100MG
m-C-tri(CH2-PEG1-NHS ester) is a non-cleavable, single-unit polyethylene glycol (PEG) antibody-drug conjugate (ADC) linker bearing an N-hydroxysuccinimide (NHS) ester for amine coupling. Supplied as a solid for research use, it provides a stable, non-cleavable spacer that improves solubility and facilitates conjugation.
  • Non-cleavable single-unit PEG linker for stable spacing
  • NHS ester functionality for efficient amine conjugation
  • Improves payload solubility
  • Supplied as a solid with ~97.0% purity (NMR)
  • Available in multiple sample sizes for research (50 mg-1 g)