Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
2,5-Thiophenedicarboxylic acid
Spécifications
Spécifications
| CAS | 4282-31-9 |
| Melting Point | >300°C (lit.) |
| Molecular Formula | C6H4O4S |
| Linear Formula | C6H4O4S |
| MDL Number | MFCD00016896 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 172.16 |
| Percent Purity | 99% |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'