Learn More
Cayman Chemical 2 5DIoxopyrrolIdIn1yl 4 p 1g
SDP
Catalog No. 502946246
Encompass_Preferred
An amine-reactive ester; has been used to coat carbon nanotube-based biosensors for the capture of antibodies and proteins
Catalog No. 50-294-6246 Supplier Cayman Chemical Supplier No. 346661 G
An amine-reactive ester; has been used to coat carbon nanotube-based biosensors for the capture of antibodies and proteins