Learn More
Cary Company 2.5 Gallon (10L) Natural Plastic Tight Head Container, Tamper Evident 70mm (6TPI), No Vent Stem
SDP
Catalog No. NC3232889
Encompass
UN Rated 2.5 Gallon (10 Liter) Rectangular Natural HDPE Plastic Tight Head Container, Tamper Evident 70mm SC-76RTE (6TPI), Unvented. 100 PER CS. The 57WSAN is a UN Rated (UN3H1/Y1.8/100) 2.5 gallon (10 liter) natural high density polyethylene (HDPE) plastic rectangular tight head container with a tamper evident 70mm (Buttress 6TPI) neck finish. This pail comes with a fixed handle. Rectangular plastic tight head containers are ideal for storing paints, coatings, adhesives, mastics, additives, agricultural chemicals, viscous products and other applications.
Catalog No. NC3232889 Supplier Cary Company Supplier No. 57WSANCS
UN Rated 2.5 Gallon (10 Liter) Rectangular Natural HDPE Plastic Tight Head Container, Tamper Evident 70mm SC-76RTE (6TPI), Unvented. 100 PER CS. The 57WSAN is a UN Rated (UN3H1/Y1.8/100) 2.5 gallon (10 liter) natural high density polyethylene (HDPE) plastic rectangular tight head container with a tamper evident 70mm (Buttress 6TPI) neck finish. This pail comes with a fixed handle. Rectangular plastic tight head containers are ideal for storing paints, coatings, adhesives, mastics, additives, agricultural chemicals, viscous products and other applications.