Learn More
Hunter Lab Glass Sample Cup
SDP
Catalog No. NC9032733
Encompass
2.5 inch (64mm) Optically clear glass cup for sample presentation of liquids, powders, granules and pellets.
This item is not returnable. View return policy
Catalog No. NC9032733 Supplier Hunter Lab Supplier No. 04720900
2.5 inch (64mm) Optically clear glass cup for sample presentation of liquids, powders, granules and pellets.