Learn More
Hach Company 2.5 mg/L NH4-N Standard Solution for Amtax sc (2000 mL)
Supplier:  Hach Company 2514754
Encompass
This ready-to-use standard solution contains 2.5 mg/L ammonium (NH4-N) in a 2,000 mL bottle and is intended for calibration and quality control of ammonia/ammonium analyzers.
  • Ready-to-use 2,000 mL calibration standard.
  • Concentration of 2.5 mg/L NH4-N.
  • Supplied in a sealed bottle for stability and handling.
  • Intended for calibration and routine quality control of analyzers measuring ammonium/ammonia.
  • Available for recurring orders to support regular maintenance schedules.
Catalog No. NC3659969