Learn More
Gerstel GERSTEL 2.5 mL Gastight Syringe, Replaceable Needle, Low Carry-Over, Durable Construction, 2.5 mL
SDP
Catalog No. NC1782460
Encompass
The GERSTEL 2.5 mL Gastight Syringe, designed for high-precision automated headspace analysis, ensures excellent accuracy and reliability. Its unique gastight design with a replaceable needle and active plunger tip minimizes sample carry-over, making it ideal for demanding laboratory applications. The rugged construction enhances resilience to solvents, while features like an improved needle connection reduce contamination risk, setting it apart from competitors.
Catalog No. NC1782460 Supplier Gerstel Supplier No. 10010600100
The GERSTEL 2.5 mL Gastight Syringe, designed for high-precision automated headspace analysis, ensures excellent accuracy and reliability. Its unique gastight design with a replaceable needle and active plunger tip minimizes sample carry-over, making it ideal for demanding laboratory applications. The rugged construction enhances resilience to solvents, while features like an improved needle connection reduce contamination risk, setting it apart from competitors.