Learn More
Trajan Scientific L-MARK Brand 2.5mL Syringe, RN, GT, Pt. 3, 4"" Needle
Catalog No. 50635275
Encompass_Preferred
L-MARK Brand 2.5mL, Syringe, PURIFIC, RN, GT, 19g, NDL Length 104mm, Pt. Style 3, Plunger Tip PTFE, Single Made in Germany.
Catalog No. 50-635-275 Supplier Trajan Scientific Supplier No. LMK.2620025
L-MARK Brand 2.5mL, Syringe, PURIFIC, RN, GT, 19g, NDL Length 104mm, Pt. Style 3, Plunger Tip PTFE, Single Made in Germany.