Learn More
Leica Biosystems 2.5ml NCL-DYS1
Supplier:  Leica Biosystems DYS1A
Encompass
Dystrophin AntibodiesDystrophin is the 427kD protein product of the DMD gene located on the X chromosome at position Xp21.Analyte Specific Reagent. Analytical and performance characteristics are not established.
Catalog No. 50-255-2270