Learn More
Gerstel GERSTEL 2.5 mL Smart TriStar Syringe, Gas-Tight, Replaceable Needle, PFAS-Free, 2.5 mL
SDP
Catalog No. NC2959172
Encompass
The GERSTEL 2.5 mL Smart TriStar HS Syringe is designed for precise and reliable sample introduction in robotic systems. Its gas-tight construction and replaceable needle enhance analytical performance, making it ideal for headspace applications. Featuring 'Smart' technology for error-free operation and traceability, it operates effectively at temperatures up to 150°C, setting it apart from competitors in the market.
Catalog No. NC2959172 Supplier Gerstel Supplier No. 10010630100
The GERSTEL 2.5 mL Smart TriStar HS Syringe is designed for precise and reliable sample introduction in robotic systems. Its gas-tight construction and replaceable needle enhance analytical performance, making it ideal for headspace applications. Featuring 'Smart' technology for error-free operation and traceability, it operates effectively at temperatures up to 150°C, setting it apart from competitors in the market.