Learn More
Sutter Instruments 2.5MM SQ FILAMENT 4.5ML WIDE
SDP
Catalog No. NC9198700
Encompass_Preferred
Filament for P-1000, P-97, P87, P80PC, P80C, PC-84, P-77B
Catalog No. NC9198700 Supplier Sutter Instruments Supplier No. FGFB245B
Filament for P-1000, P-97, P87, P80PC, P80C, PC-84, P-77B