Learn More
Medchemexpress LLC 2-((6-(4-chlorophenyl)-1-methyl-4H-triazolo[4,3-a]azepin-4-yl)methyl)-5-methyl-1,3,4-oxadiazole | 1588521-78-1 | 99.8% | 403.86 g/mol | C22H18ClN5O | 10MG
SDP
Catalog No. 5000237008
Encompass_Preferred
BET-BAY 002 is a research-grade small-molecule BET bromodomain inhibitor used in epigenetic and oncology research. It is supplied as a white to off-white solid (CAS 1588521-78-1) with a molecular weight of 403.86 g/mol and reported high purity for reliable experimental use. The compound dissolves readily in DMSO and has recommended storage conditions for long-term stability.
Catalog No. 50-002-37008 Supplier Medchemexpress LLC Supplier No. HY1242110MG
BET-BAY 002 is a research-grade small-molecule BET bromodomain inhibitor used in epigenetic and oncology research. It is supplied as a white to off-white solid (CAS 1588521-78-1) with a molecular weight of 403.86 g/mol and reported high purity for reliable experimental use. The compound dissolves readily in DMSO and has recommended storage conditions for long-term stability.