Learn More
Medchemexpress LLC 2-[(6-amino-9H-purin-9-yl)methyl]-5-methyl-3-(2-methylphenyl)-4(3H)-quinazolinone | 371242-69-2 | MFCD09970845 | >98.0% | 397.4 g/mol | C22H19N7O | 1 ML
SDP
Catalog No. 5000211422
Encompass_Preferred
IC-87114 is a selective small-molecule inhibitor of PI3Kδ (p110δ) used in research to probe PI3K signaling and immune-cell functions. It displays reported potency (IC50 ≈ 0.5 μM) and is supplied as a 10 mM solution in a 1 mL vial for analytical and experimental use.
  • Selective inhibition of PI3Kδ (p110δ).
  • Potent activity with IC50 ~0.5 μM.
  • Supplied as a 10 mM solution in a 1 mL vial.
  • High purity suitable for research use.
  • Applicable to immunology and cell-signaling studies.
Catalog No. 50-002-11422 Supplier Medchemexpress LLC Supplier No. HY1011010MM 1ML
IC-87114 is a selective small-molecule inhibitor of PI3Kδ (p110δ) used in research to probe PI3K signaling and immune-cell functions. It displays reported potency (IC50 ≈ 0.5 μM) and is supplied as a 10 mM solution in a 1 mL vial for analytical and experimental use.
  • Selective inhibition of PI3Kδ (p110δ).
  • Potent activity with IC50 ~0.5 μM.
  • Supplied as a 10 mM solution in a 1 mL vial.
  • High purity suitable for research use.
  • Applicable to immunology and cell-signaling studies.