Learn More
Cayman Chemical 2- 6-Aminohexylthioadenosine
SDP
Catalog No. 502918717
Encompass_Preferred
An aminohexylthiol-substituted derivative of ATP; has been used in the synthesis of biotinylated derivatives of ATP
Catalog No. 50-291-8717 Supplier Cayman Chemical Supplier No. 38545600 UG
An aminohexylthiol-substituted derivative of ATP; has been used in the synthesis of biotinylated derivatives of ATP