Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(6-azaspiro[2.5]octan-6-yl)-4-(2-hydroxyethylsulfonylamino)-N-[6-[(2R)-2-methylmorpholin-4-yl]pyrid... | 2410796-89-1 | 99.2% | C26H35N5O5S | 10MG
SDP

Catalog No. 5000259902
Encompass_Preferred

AM-5308 is a small-molecule inhibitor of KIF18A used as a research reagent to probe mitotic checkpoint signaling, chromosome segregation, and spindle assembly. It has reported antitumor activity in preclinical studies and is supplied as a high-purity powder with defined solubility and storage recommendations.

  • High purity (99.2%).
  • Molecular formula C26H35N5O5S; molecular weight 529.65 g/mol.
  • Soluble in DMSO at 50 mg/mL; may require sonication.
  • Recommended storage: powder -20°C for long-term; in solvent -80°C for up to 6 months.
  • Potent inhibitor of KIF18A demonstrated in biochemical and cellular studies.
  • Intended for preclinical research into mitotic regulation and antitumor mechanisms.

Catalog No. 50-002-59902 Supplier Medchemexpress LLC Supplier No. HY14489410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AM-5308 is a small-molecule inhibitor of KIF18A used as a research reagent to probe mitotic checkpoint signaling, chromosome segregation, and spindle assembly. It has reported antitumor activity in preclinical studies and is supplied as a high-purity powder with defined solubility and storage recommendations.

  • High purity (99.2%).
  • Molecular formula C26H35N5O5S; molecular weight 529.65 g/mol.
  • Soluble in DMSO at 50 mg/mL; may require sonication.
  • Recommended storage: powder -20°C for long-term; in solvent -80°C for up to 6 months.
  • Potent inhibitor of KIF18A demonstrated in biochemical and cellular studies.
  • Intended for preclinical research into mitotic regulation and antitumor mechanisms.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.