Learn More
Medchemexpress LLC 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, hydrochloride (1:2), (6S)- | 104632-25-9 | 98.0% | 284.25 | 5 MG
SDP
Catalog No. 5000384558
Encompass_Preferred
Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist. It is used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective dopamine D2-type receptor agonist
  • Blood-brain barrier penetrant
  • Used for Parkinson's disease research
  • Used for restless legs syndrome research
  • Has Ki values for D2-type, D2, D3, and D4 receptors
Catalog No. 50-003-84558 Supplier Medchemexpress LLC Supplier No. HY173555MG
Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist. It is used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective dopamine D2-type receptor agonist
  • Blood-brain barrier penetrant
  • Used for Parkinson's disease research
  • Used for restless legs syndrome research
  • Has Ki values for D2-type, D2, D3, and D4 receptors