Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(trimethylstannyl)-4,8-bis[(2-n-octyldodecyl)oxy]benzo[1,2-b:4,5-b']dithiophene 95.0+%, TCI America™
SDP

Catalog No. B4488200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4488200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4488200MG Supplier TCI America Supplier No. B4488200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1320201-22-6
Molecular Formula C56H102O2S2Sn2
Molecular Weight (g/mol) 1108.97
MDL Number MFCD29472510
InChI Key AZENTBKKKXIGMB-UHFFFAOYNA-N
PubChem CID 129852992
IUPAC Name {2,8-bis[(2-octyldodecyl)oxy]-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl}trimethylstannane
SMILES CCCCCCCCCCC(CCCCCCCC)COC1=C2C=C(SC2=C(OCC(CCCCCCCC)CCCCCCCCCC)C2=C1SC(=C2)[Sn](C)(C)C)[Sn](C)(C)C

Specifications

Quantity 200 mg
UN Number 3146
Formula Weight 1108.97
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(trimethylstannyl)-4,8-bis[(2-n-octyldodecyl)oxy]benzo[1,2-b:4,5-b′]dithiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.