Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diaminopimelic acid, 96%

Catalog No. AAB2239106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2239106 5 g
AAB2239103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2239106 Supplier Thermo Scientific Chemicals Supplier No. B2239106
Only null left
Add to Cart

CAS: 583-93-7 | C7H14N2O4 | 190.199 g/mol

2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.

Solubility
Soluble in water, diluted acids and alkali.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-93-7
Molecular Formula C7H14N2O4
Molecular Weight (g/mol) 190.199
MDL Number MFCD00002637
InChI Key GMKMEZVLHJARHF-UHFFFAOYSA-N
Synonym 2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm
PubChem CID 865
ChEBI CHEBI:23673
IUPAC Name 2,6-diaminoheptanedioic acid
SMILES C(CC(C(=O)O)N)CC(C(=O)O)N

Specifications

Melting Point >300°C
Assay Percent Range 0.98
Quantity 5 g
Solubility Information Soluble in water,diluted acids and alkali.
Formula Weight 190.2
Chemical Name or Material 2, 6-Diaminopimelic Acid
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-524-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.