Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diaminopyridine 98.0+%, TCI America™
SDP

Catalog No. D115425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D115425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D115425G Supplier TCI America Supplier No. D115425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory reagent with minimum 98.0% by GC and titration is supplied as a white to brown solid.

Chemical Identifiers

CAS 141-86-6
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD00006329
InChI Key VHNQIURBCCNWDN-UHFFFAOYSA-N
Synonym 2,6-diaminopyridine, 2,6-pyridinediamine, 2,6-diamino pyridine, pyridine, 2,6-diamino, dap amine, pyridine-2,6-diyldiamine, unii-k89ab8ckg6, ccris 6682, k89ab8ckg6, dsstox_cid_20127
PubChem CID 8861
IUPAC Name pyridine-2,6-diamine
SMILES C1=CC(=NC(=C1)N)N

Specifications

Melting Point 122°C
Boiling Point 285°C
Quantity 25 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diaminopyridine
EINECSNumber (5)-0726
RTECSNumber US7570000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.