Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diaminotoluene, 98%

Catalog No. AC164680250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164680250 25 g Glass bottle
AC164680050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164680250 Supplier Thermo Scientific Chemicals Supplier No. 164680250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 823-40-5 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-40-5
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007800
InChI Key RLYCRLGLCUXUPO-UHFFFAOYSA-N
Synonym 2,6-diaminotoluene, 2,6-toluenediamine, 1,3-benzenediamine, 2-methyl, toluene-2,6-diamine, 2,6-tolylenediamine, 2,6-toluylenediamine, 2,6-diamino-1-methylbenzene, 2-methyl-1,3-benzenediamine, 2-methyl-1,3-phenylenediamine, 2-methyl-m-phenylenediamine
PubChem CID 13205
ChEBI CHEBI:76288
IUPAC Name 2-methylbenzene-1,3-diamine
SMILES CC1=C(N)C=CC=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 102.0°C to 107.0°C
Color Black or Brown to Gray
Boiling Point 289.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NH2)2
Quantity 25 g
Beilstein 13,148
Solubility Information Solubility in water: 60g/L water (15°C).
Formula Weight 122.17
Percent Purity 98%
Physical Form Pellets, Powder or Chunks
Chemical Name or Material 2, 6-Diaminotoluene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if y
EINECSNumber 212-513-9
RTECSNumber XS9750000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.