Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromo-1,4-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. D43841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D43841G Supplier TCI America Supplier No. D43841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29213-03-4
Molecular Formula C6H6Br2N2
Molecular Weight (g/mol) 265.94
MDL Number MFCD02575406
InChI Key NIXNGYGWQMXMCA-UHFFFAOYSA-N
Synonym 2,5-Diamino-1,3-dibromobenzene
PubChem CID 653341
IUPAC Name 2,6-dibromobenzene-1,4-diamine
SMILES NC1=CC(Br)=C(N)C(Br)=C1

Specifications

Melting Point 138°C
Quantity 1 g
Formula Weight 265.94
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-1,4-phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.