Learn More
Indofine Chemical 2,6-Dibromo-4-(Trif, 95%, 5 Gm
SDP
Catalog No. 50910520
Encompass_Preferred
2,6-DIBROMO-4-(TRIFLUOROMETHYL)ANILINE 5 GM, Fluoro Organics, 4-Amino-3,5-dibromobenzotrifluoride, C7H4Br2F3N, MW: 318.93, 95+%, 72678-19-4, MFCD00068181
Catalog No. 50-910-520 Supplier Indofine Chemical Supplier No. 09767 5 GM
2,6-DIBROMO-4-(TRIFLUOROMETHYL)ANILINE 5 GM, Fluoro Organics, 4-Amino-3,5-dibromobenzotrifluoride, C7H4Br2F3N, MW: 318.93, 95+%, 72678-19-4, MFCD00068181