Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene 98.0+%, TCI America™
SDP

Catalog No. D37991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37991G 1 g
D3799200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37991G Supplier TCI America Supplier No. D37991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67061-69-2
Molecular Formula C8H2Br2S3
Molecular Weight (g/mol) 354.092
InChI Key UTKPZZGBROTDKR-UHFFFAOYSA-N
PubChem CID 11382649
IUPAC Name 2,6-dibromodithieno[2,3-a
SMILES C1=C(SC2=C1SC3=C2SC(=C3)Br)Br

Specifications

Melting Point 171°C
Quantity 1 g
Formula Weight 354.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromodithieno[3,2-b:2′,3′-d]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.