Learn More
Apexbio Technology LLC 2-6-DIBROMOPHENOL-500MG
SDP
Catalog No. 5000568735
Encompass_Preferred
2 6-Dibromophenol (CAS No 608-33-3) is a halogenated phenolic compound frequently studied in the field of environmental toxicology and pharmacology Originating naturally from marine organisms it has been shown to modulate specific biochemical pathways including enzyme inhibition and receptor interactions Its in vitro activity is often observed in the nanomolar to low micromolar range reflecting its potency in various experimental settings The compound is commonly used in cell models to evaluate its potential effects and mechanisms of toxicity or therapeutic action In addition it serves as a reference compound in animal studies to investigate its pharmacokinetics and pharmacodynamics It is also employed in drug screening assays to explore its interactions with molecular targets Experimental concentrations typically depend on the specific research design and studies may vary in their focus on acute or chronic effects within controlled environments
Catalog No. 50-005-68735 Supplier Apexbio Technology LLC Supplier No. C6618500
2 6-Dibromophenol (CAS No 608-33-3) is a halogenated phenolic compound frequently studied in the field of environmental toxicology and pharmacology Originating naturally from marine organisms it has been shown to modulate specific biochemical pathways including enzyme inhibition and receptor interactions Its in vitro activity is often observed in the nanomolar to low micromolar range reflecting its potency in various experimental settings The compound is commonly used in cell models to evaluate its potential effects and mechanisms of toxicity or therapeutic action In addition it serves as a reference compound in animal studies to investigate its pharmacokinetics and pharmacodynamics It is also employed in drug screening assays to explore its interactions with molecular targets Experimental concentrations typically depend on the specific research design and studies may vary in their focus on acute or chronic effects within controlled environments