Learn More
Medchemexpress LLC 2,6-dibromophenol | 608-33-3 | MFCD00002152 | 100g
SDP
Catalog No. 5000265938
Encompass_Preferred
2 6-Dibromophenol (2 6-DBP) is a weak inhibitor targeting voltage-gated Ca2 channels 2 6-Dibromophenol has no significant effect on potassium-induced calcium elevation in PC12 cells 2 6-Dibromophenol shows a 48 h EC50 of 2 78 mg/L for Daphnia magna and a 96 h EC50 of 9 90 mg/L for algae (Scenedesmus quadricauda)[1][2]
Catalog No. 50-002-65938 Supplier Medchemexpress LLC Supplier No. HYY1667100G
2 6-Dibromophenol (2 6-DBP) is a weak inhibitor targeting voltage-gated Ca2 channels 2 6-Dibromophenol has no significant effect on potassium-induced calcium elevation in PC12 cells 2 6-Dibromophenol shows a 48 h EC50 of 2 78 mg/L for Daphnia magna and a 96 h EC50 of 9 90 mg/L for algae (Scenedesmus quadricauda)[1][2]