Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-3-cyano-4-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. D33465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33465G 5 g
D334625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33465G Supplier TCI America Supplier No. D33465G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 875-35-4
Molecular Formula C7H4Cl2N2
Molecular Weight (g/mol) 187.023
MDL Number MFCD00052633
InChI Key LSPMHHJCDSFAAY-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-methylnicotinonitrile, 2,6-dichloro-3-cyano-4-methylpyridine, 3-cyano-2,6-dichloro-4-methylpyridine, 3-cyano-2,6-dichloro-4-methyl pyridine, 3-pyridinecarbonitrile, 2,6-dichloro-4-methyl, 3-cyano-4-methyl-2,6-dichloropyridine, 2,6-dichloro-4-methyl-3-cyanopyridine, 2,6-dichloro-4-methylnicotinonitrile 3-cyano-2,6-dichloro-4-methylpyridine, pubchem15375, acmc-209qo9
PubChem CID 70131
IUPAC Name 2,6-dichloro-4-methylpyridine-3-carbonitrile
SMILES CC1=CC(=NC(=C1C#N)Cl)Cl

Specifications

Melting Point 111°C
Boiling Point 118°C
Quantity 5 g
UN Number 3439
Formula Weight 187.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-3-cyano-4-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.